Please login to your online account to display your discounted pricing

Milrinone, Tocris

PDE3 inhibitor

$221.01 - $904.70

Chemical Identifiers

Chemical Name or Material Milrinone
Assay Percent Range >98%
Molecular Formula C12H9N3O
Formula Weight 211.22
Synonym 1, 6-Dihydro-2-methyl-6-oxo-(3, 4'-bipyridine)-5-carbonitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

150450

 
r & d systems
RDS150450
50mg Each for $904.70

150410

 
r & d systems
RDS150410
10mg Each for $221.01
Description & Specifications

Specifications

Formula Weight 211.22
Quantity 50mg
Chemical Name or Material Milrinone
CAS 78415-72-2
Assay Percent Range >98%
Synonym 1, 6-Dihydro-2-methyl-6-oxo-(3, 4'-bipyridine)-5-carbonitrile
Molecular Formula C12H9N3O
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C