Please login to your online account to display your discounted pricing

Milnacipran hydrochloride, Tocris

5-HT and noradrenalin re-uptake inhibitor (SNRI)

$126.32 - $534.65

Chemical Identifiers

Chemical Name or Material Milnacipran hydrochloride
Assay Percent Range >99%
Molecular Formula C15H22N2O·HCl
Formula Weight 282.81
Synonym (1R™, 2S™)-2-(Aminomethyl)-N, N-diethyl-1-phenylcyclopropanecarboxamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

328650

 
r & d systems
RDS328650
50mg Each for $534.65

328610

 
r & d systems
RDS328610
10mg Each for $126.32
Description & Specifications

Specifications

Formula Weight 282.81
Quantity 50mg
Chemical Name or Material Milnacipran hydrochloride
Molecular Formula C15H22N2O·HCl
Assay Percent Range >99%
CAS 101152-94-7
Synonym (1R™, 2S™)-2-(Aminomethyl)-N, N-diethyl-1-phenylcyclopropanecarboxamide hydrochloride
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C