Learn More
MilliporeSigma Milli-Q™ SP 240 Prefilter Module
Description
- The Milli-Q™ SP 240 Prefilter Module pretreats potable tap water to feed the pure RO water production station.
- This prefilter module is recommended to boost the performance of the RO water production station of the Milli-Q™ SQ 2Series of water purification systems.
Specifications
Specifications
| Feedwater Source | Potable Tap Water |
| Flow Rate | 42 L/hr. |
| Type | Ultrapure Water Purification System |