MilliporeSigma™ Millex™ Syringe Filter, Durapore™ (PVDF), 0.22 μm
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
$240.00 - $2,420.00
Specifications
Disposable Yes
Pore Size 0.22 μm
Sterility Non-sterile
For Use With (Application) Analytical sample prep
Inlet Style Female Luer-Lok™
View More Specs
Products 7
Catalog Number Mfr. No. Hold up Volume (After Air Purge) Length (Metric) Outlet Style Quantity Price Quantity  
Catalog Number Mfr. No. Hold up Volume (After Air Purge) Length (Metric) Outlet Style Quantity Price Quantity  
SLGVX13NK
View Documents Promotion Details
MilliporeSigma™
SLGVX13NK
≤15 µL 21 mm Male Luer slip 1000Pack
Pack of 1000 Each for $2,205.01
SLGVX13NL
View Documents Promotion Details
MilliporeSigma™
SLGVX13NL
≤15 µL 21 mm Male Luer slip 100Pack
Pack of 100 Each for $352.00
SLGVX13TL
View Documents Promotion Details
MilliporeSigma™
SLGVX13TL
≤15 µL 48 mm Tube outlet 100Pack
Pack of 100 Each for $370.00
SLGV033NB
View Documents Promotion Details
MilliporeSigma™
SLGV033NB
≤80 µL 25 mm Male Luer slip Pk. of 250
Pack of 250 Each for $914.50
SLGV033NS
View Documents Promotion Details
MilliporeSigma™
SLGV033NS
≤80 µL 27 mm Male Luer slip Pk. of 50
Pack of 50 Each for $240.00
SLGVR04NK
View Documents Promotion Details
MilliporeSigma™
SLGVR04NK
≤10 µL 19.7 mm Male Luer slip, stepped 1000Pack
Pack of 1000 Each for $2,420.00
SLGVR04NL
View Documents Promotion Details
MilliporeSigma™
SLGVR04NL
≤10 µL 19.7 mm Male Luer slip, stepped 100Pack
Each for $250.49
Description
Description
For removing particulates, reducing signal-to-noise ratios and maintaining flat baselines
Overview of Non-Sterile Millex™ Syringe Filters with Durapore™ (PVDF) Membrane
  • Low protein binding to minimize interaction with your sample and maximize recovery
  • Compatible with aqueous and mild organic solutions
  • Housing materials are either polypropylene or high density polyethylene (HDPE), offering low levels of extractables and broad chemical compatibility
Specifications
Specifications
Yes
Non-sterile
Female Luer-Lok™
0.22 μm
Analytical sample prep
Syringe Filter