Honeywell™ Miller™ TranScendor™ Cable Climbing Systems
Permanently installed, flexible cable system provides continuous fall protection for climbing fixed ladders
Manufacturer:  Honeywell™ TRS400FT
 View more versions of this product
Catalog No. 19-074-565
Includes: Top bracket assembly with shock absorber, a bottom bracket assembly with lifeline tensioner, 3/8 in. (9.5mm) galvanized steel cable wire rope lifeline, all necessary hardware, cable guides (when applicable) and instruction manual
Description
Description
  • Quick, easy, do-it-yourself installation and inspection
  • Lifeline tensioner prevents ladder rung deformation and indicates when the lifeline is properly adjusted
  • Quick-link design for trouble-free cable installation
  • Cable guides are fully convertible to accommodate low and high wind conditions increasing versatility
  • Provides fall protection for up to four workers simultaneously
  • Durable cable sleeve locks instantly in the event of a fall and attaches easily to the cable
Specifications
Specifications
OSHA, ANSI, CSA
Manual pass-through
121.92 m
Ideal for agriculture, construction, manufacturing, municipal services, Oil and gas, Steel and Metals, utilities
400 ft.