Honeywell™ Miller™ TranScendor™ Cable Climbing Systems
Permanently installed, flexible cable system provides continuous fall protection for climbing fixed ladders
Manufacturer: Honeywell™ TRS50FT
Includes: Top bracket assembly with shock absorber, a bottom bracket assembly with lifeline tensioner, 3/8 in. (9.5mm) galvanized steel cable wire rope lifeline, all necessary hardware, cable guides (when applicable) and instruction manual
Description
- Quick, easy, do-it-yourself installation and inspection
- Lifeline tensioner prevents ladder rung deformation and indicates when the lifeline is properly adjusted
- Quick-link design for trouble-free cable installation
- Cable guides are fully convertible to accommodate low and high wind conditions increasing versatility
- Provides fall protection for up to four workers simultaneously
- Durable cable sleeve locks instantly in the event of a fall and attaches easily to the cable
Specifications
OSHA, ANSI, CSA | |
Manual pass-through | |
1524 cm |
Ideal for agriculture, construction, manufacturing, municipal services, Oil and gas, Steel and Metals, utilities | |
50 ft. |