Honeywell™ Miller™ Roofing Kit Accessories
Carrying bag for easy organization and transportation of Miller Roof Anchor Kits.
Manufacturer: Honeywell™ 8477HYL
Description
Miller Roof Anchor Kits are easy to use and install on steep-pitched or flat-surface roofs. The kits include: 2 reusable roof anchors, steel twist-lock carabiner, nylon rope lifeline, manual stainless steel rope grab with 2 ft. (0.6m) webbing lanyard with shock-absorber and DuraFlex stretchable harness. Kits meet all applicable OSHA, ANSI and CSA standards.
Specifications
Roofing kit | |
Yellow | |
Carrying bag |