missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Honeywell™ Miller™ Manyard™ HP™ Shock-Absorbing Lanyards

Reduce the force of a fall by up to 80%

Manufacturer:  Honeywell™ 231TWRSZ76FTGNC**

Catalog No. 19066223



Description

Description

  • Specially woven shock-absorbing inner core smoothly expands up to 42 in. (1.1m) to reduce fall arrest
  • Heavy-duty tubular outer jacket serves as a back-up web lanyard
  • Warning flag provides visual proof unit has been involved in a fall and must be removed from service
  • Locking snap-hook harness connections
  • Single-leg style
  • Hi-vis green

Order Info

Length: 6 ft. (1.8m).

Compliance

Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI™ A10.32a nd ANSI Z359.1; CSA Z259.11 requirements.

Specifications

Specifications

Meets ANSI Z359-2007, and all applicable OSHA, ANSI and CSA standards.
Double-Leg
6 ft.
Webbing
Locking Rebar Hooks
Hi-vis Green
1.8 m
SDS
Documents