Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Milk Fat Globule 1, Human (HMFG1) Ab-1, Mouse Monoclonal Antibody


Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Milk Fat Globule 1, Human (HMFG1) Ab-1, Mouse Monoclonal Antibody.

Manufacturer: thermo scientific™ lab vision™  MS512P1ABX

 View more versions of this product

Catalog No. MS512P1ABX

  • / Each
 

Description & Specifications

Specifications

Antigen Milk Fat Globule 1, Human (HMFG1) Ab-1
Immunogen Delipidated human milk fat globule membrane preparation
Host Species Mouse
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Human
Format Concentrated
Quantity 100µL
Clone EDM45
Isotype IgG2a kappa
Formulation Ab without BSA/Azide (Frozen)
Research Discipline Cancer and Tumor Biology
Type Primary
Classification Monoclonal
Concentration 1mg/mL
Conjugate Unlabeled

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with the Thermo Scientific™ Milk Fat Globule 1, Human (HMFG1) Ab-1, Mouse Monoclonal Antibody.

Human milk fat globule membrane protein (HMFG1) is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as on the outer surface of the human milk fat globule. HMFG1 is considered as a differentiation marker. It is useful as specific breast epithelial marker in normal and neoplastic mammary development.

Host Species: Mouse

Clone: EDM45

Isotype: IgG1/κ

Species Reactivity: Human. Does not react with rat. Others not known.

Epitope: Not determined

Immunogen: Delipidated human milk fat globule membrane preparation

Molecular Weight: 40-45kDa

Positive Control: SKBR-3 cells; Breast, colon, or lung tissues

Cellular Localization: Membrane and cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO