missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Hoefer™ Mighty Small™ II Mini Vertical Electrophoresis Systems: PROMO

10% off promotion on the Mighty Small™ II Mini Vertical Electrophoresis Systems

Manufacturer:  Hoefer™ SE25010A75PR

 View more versions of this product

Catalog No. 03500110PM



Description

Description

  • Run two gels at once
  • Produce high-resolution separations in as little as 45 minutes with no band “smiling”
  • Use alumina plates (included with specified models) for efficient cooling
  • Buffer volume: 300mL

Applications:

Ideal for uniform and gradient acrylamide, agarose, IEF, field inversion agarose and 2-D electrophoresis

Specifications

Specifications

500mA
16cm
Lower buffer chamber, upper buffer chamber/cooling core, safety lid with high voltage leads, casting cradle with sealing gasket set, casting clamp assemblies, cams, glass plates, rectangular, 10 x 8cm, alumina plates, notched, 10 x 8cm, spring clamps, com
45°C
0.2 in.
500V
10.14 oz.
3.14 in.
3.93 in.
6.29 in.
6.49 in.
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
6.29 in.
Up to 80%
113°F
15° to 30°C
0.07cm
12w
300mL
8cm
10cm
16cm
16.5cm
Documents