Please login to your online account to display your discounted pricing

Middle School Simple Machines Bulletin Board Set


A colorful and informative science-themed bulletin board set.

Manufacturer: fisher science education  CD-410031

 View more versions of this product

Catalog No. S02425

  • / Each
 

Description & Specifications

Specifications

Category Physics
Product Type Bulletin Board Set
Grade 5 to 8
Includes 5 Charts
For Use With (Application) Colorful and informative science themed bulletin board set