missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Microtiter Tube Plug Strip
Seal 1.2mL microtiter tubes tightly for storage using these polyethylene plug strips.
$76.31 - $76.31
Specifications
Description | 1.2mL Microtiter Tube 8-cap plug strip |
---|---|
Autoclavable | Not Autoclavable |
Color | Natural |
Material | LDPE |
Packaging | 10 sealed bags of twelve 8 cap strips/unit, 5 units/case |
Catalog Number | Mfr. No. | Description | Sterility | Color | Packaging | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Sterility | Color | Packaging | Price | Quantity | ||
21-403-159
|
Thermo Scientific™
3425 |
1.2mL Microtiter Tube 8-cap plug strip | Sterile | Natural | 10 sealed bags of twelve 8 cap strips/unit, 5 units/case |
Pack of 120 Each for $76.31
Case of 5 Pack for $381.55 |
|
Description
Seal 1.2mL microtiter tubes tightly for storage by using Thermo Scientific™ Microtiter Tube Plug Strips.
- LDPE non-autoclavable
- Eight-plug strips can be easily cut apart for individual samples
- Available sterile and nonsterile
Specifications
1.2mL Microtiter Tube 8-cap plug strip | |
Natural | |
10 sealed bags of twelve 8 cap strips/unit, 5 units/case |
Not Autoclavable | |
LDPE |