Micronova™ Vial Tool
$171.95 - $1,445.69
Specifications
Certifications/Compliance ISO 9001 : 2008
For Use With (Equipment) SSU series of extendible and fixed length handle
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) Autoclavable Product Type Price Quantity  
Catalog Number Mfr. No. For Use With (Application) Autoclavable Product Type Price Quantity  
19-435-302 Micronova™
VTC68IR
Minimize risk of scratching Stainless Steel surfaces Yes Irradiated cover Case of 50 Each for $1,445.69
19-435-300 Micronova™
QDPVT08P
Safely remove tipped or broken vials from lyopholizers and other cabinets Yes Vial tool adapter Each for $171.95
Specifications Description & Specifications
Specifications
ISO 9001 : 2008
SSU series of extendible and fixed length handle