Micronova™ T-Mop™ MicroFiber™ Slipcover
Low profile mop can clean most surfaces in a cleanroom
$30.33 - $31.94
Specifications
Autoclavable Yes
Material MicroFiber
Product Type Mop Cover
For Use With (Application) Cleaning floors, walls and ceilings
Includes Thick foam
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Size Width (English) Price Quantity  
Catalog Number Mfr. No. Size Width (English) Price Quantity  
19-142-641 Micronova™
TAA1218
45.72cm (18 in.) 18 in. Each for $30.33
19-142-640 Micronova™
TAA1214
35.6cm (14 in.) 14 in. Each for $31.94
Case of 12 Each for $359.76
Specifications
Description
  • T-shaped mop has a wide surface which serves as a perfect device for applying disinfectants in a sterile environment
  • With a 180° swivel, the mop is ideal for cleaning under equipment or for applying disinfectants or detergents to large surface areas
  • Smaller adapter heads can be used for cleaning surfaces and awkward angles on shelving and in narrow chambers
Description & Specifications
Specifications
Yes
Mop Cover
Thick foam
MicroFiber
Cleaning floors, walls and ceilings