Micronova™ T-Mop™ MicroFiber™ Cover
The wide surface serves as a perfect device for applying disinfectants in a sterile environment
$21.11 - $355.66
Specifications
Autoclavable Yes
Material MicroFiber
Product Type Mop Cover
Sterile Yes
For Use With (Application) Ideal for general cleaning and applying disinfectants in sterile environments
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Size Width (English) Price Quantity  
Catalog Number Mfr. No. Size Width (English) Price Quantity  
19-142-646 Micronova™
TAA14IR
35.6cm (14 in.) (Gamma) 14 in. Case of 12 Each for $355.66
19-142-630 Micronova™
TAA10
25.4cm (10 in.) 10 in. Each for $21.11
Case of 12 Each for $237.72
19-142-631 Micronova™
TAA14
35.6cm (14 in.) 14 in. Each for $23.02
Case of 12 Each for $259.32
19-142-632 Micronova™
TAA18
45.72cm (18 in.) 18 in. Each for $24.27
Case of 12 Each for $273.36
Specifications
Description
  • This low profile mop can be used to clean most surfaces in a cleanroom
  • With a 180° swivel, the mop is ideal for cleaning under equipment or for applying disinfectants or detergents to large surface areas
  • Smaller adapter heads can be used for cleaning surfaces and awkward angles on shelving and in narrow chambers
Description & Specifications
Specifications
Yes
Mop Cover
Ideal for general cleaning and applying disinfectants in sterile environments
MicroFiber
Yes
T-Mop™ mop