Micronova™ T-Mop™ Cleanroom Mops: Adapters
Low-profile, wide-surface mop
$162.12 - $163.53
Specifications
Autoclavable Yes
Material Stainless Steel, Delrin™
Product Type Quick Disconnect Adapter
Sterile Yes
For Use With (Equipment) T-Mop™ cleanroom mop
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Width (English) Price Quantity  
Catalog Number Mfr. No. Width (English) Price Quantity  
19-035-363R Micronova™
QDT18
18 in. Each for $162.12
19-035-363E Micronova™
QDT14
14 in. Each for $163.53
Specifications
Description
  • Ideal for applying disinfectants in sterile environments or for general cleaning in ISO Class 3 to 8 cleanroom environments
  • 180° swivel allows it to clean easily under equipment or in awkward corners and tight spaces
  • Autoclavable “quick-disconnect” stainless-steel/Delrin™ frame
Description & Specifications
Specifications
Yes
Quick Disconnect Adapter
T-Mop™ cleanroom mop
Stainless Steel, Delrin™
Yes