Micronova™ SnapMop™: Non-Irradiated
Can be laundered and autoclaved
$472.10 - $595.93
Specifications
Autoclavable Yes
For Use With (Application) For cleanroom use
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Width (English) Material Product Type Price Quantity  
Catalog Number Mfr. No. Includes Width (English) Material Product Type Price Quantity  
19-305-851 Micronova™
MZSM1114
With Polyurethane Foam interior 14 in. PolySorb Mop Case of 24 Each for $500.90
19-305-850 Micronova™
MZSM1118
With Polyurethane Foam interior 18 in. PolySorb Mop Case of 24 Each for $528.04
19-166-750 Micronova™
MZSM6AA18
With Polyurethane Foam interior 18 in. Microfiber Cover Case of 24 Each for $523.11
19-305-688 Micronova™
MZSM6814
With Polyurethane Foam interior 14 in. Polyester Mop Case of 24 Each for $472.10
19-045-260 Micronova™
MZSM1114NS
Without Polyurethane Foam interior 14 in. PolySorb Mop Case of 50 Each for $595.93
Specifications
Description
  • Polyester/microfiber/urethane foam
  • Laundered and autoclaved
  • Will hold up to alcohol, strong disinfectants and steam sterilization
  • Non-irradiated
Description & Specifications
Specifications
Yes
For cleanroom use