Micronova™ Slim T™ Double-Bucket Cart and Wringer
Suitable for use with SlimLine™ and T-Mop™ Series and a variety of other mop heads
$305.93 - $1,324.37
Specifications
Autoclavable Yes
Quantity Each
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. For Use With (Application) For Use With (Equipment) Material Product Type Price Quantity  
Catalog Number Mfr. No. For Use With (Application) For Use With (Equipment) Material Product Type Price Quantity  
19-121-801 Micronova™
C7*
Cleanroom and cleanroom support Slimline™ and T-Mop™ series and a variety of other mop heads Polypropylene, Stainless Steel Double Bucket Cart Each for $1,324.37
19-121-802 Micronova™
W9
Allows the operator to wring out excess fluid leaving the mop damp enough to effectively saturate the surface being cleaned SlimT™ double bucket cart, T-Mop™ or squeegee style mops, C-7 cart or B-7 series bucket Stainless Steel SlimT™ Wringer Each for $305.93
Specifications
Description
  • Simple and effective double-bucket system with casters for 360° maneuverability
  • Stainless-steel frame easily transports two 6-gal. polypropylene buckets (included)
  • Optional stainless-steel wringer fits inside bucket to squeeze excess liquid from mop, but leaves it damp enough to effectively clean
  • Bucket cart, buckets and wringer are autoclavable
Order Info
Each bucket measures 19.25 × 9.5 × 9.5 in. (48.9 × 24.1 × 24.1cm).
Description & Specifications
Specifications
Yes
Each