Micronova™ PolyGen™ Mop Heads
Economical and low-linting
$70.77 - $70.77
Specifications
Autoclavable Yes
Material Polyester
Sterile Yes
For Use With (Application) ISO class 3 to 8 cleanrooms, floor
Includes Herringbone weave in its looped strands
View More Specs
Requires: 19-035-363A Quick Disconnect MicroMop™ Adapter and extendable or single-length stainless-steel handle (sold separately).
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
19-059-163 Micronova™
PG912
PolyGen™ Mophead Each for $70.77
Specifications
Description
  • For ISO Class 3 to 8 cleanrooms
  • 100% polyester with a herringbone weave in its looped strands
  • Made to hold up to strong disinfectants and autoclaving
  • No raw edges assure low particulation
  • Absorbent enough to clean to dry finish
  • Also effective with disinfectants that need extended drying time
Description & Specifications
Specifications
Yes
Yes
Herringbone weave in its looped strands
Polyester
ISO class 3 to 8 cleanrooms, floor