missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Micronova™ Polyethylene Cleanroom Buckets

For use with SlimLine™ Cleanroom Cart System

$224.66 - $232.97

Specifications

Product Type Slimeline bucket
Autoclavable Yes
For Use With (Equipment) SlimLine or SlimT Cleanroom Cart system
Includes 3 in. casters
Material Polyethylene
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
19-065-100 Micronova™
B7DBAC
Dark Blue Each for $232.97
19-065-103 Micronova™
B7YEAC
Yellow Each for $224.66
Specifications

Description

  • With 3 in. casters
  • 6 gallons
  • Autoclavable
  • 19.25 x 9.5 x 9.5 in.

Description & Specifications

Specifications

Slimeline bucket
SlimLine or SlimT Cleanroom Cart system
Polyethylene
Yes
3 in. casters