Micronova™ PadMop™ Sponge Mop
For use in pharmaceutical production or aseptic processing areas
Manufacturer:  Micronova™ SLP618
 View more versions of this product
Catalog No. 19-130-4830
Description
Description
  • Covered sponge mop designed to apply the optimum amount of disinfectant
  • Brick-style mop fits onto SlimLine™ QDSL adapters creating a fully padded mop head for large surface areas, shelving, door jambs, and corners
  • Absorbent foam interior retains liquid well; when pressure is applied, moisture is released to fully saturate surfaces
  • Used to disinfect most large surfaces in a production area
  • Gamma irradiated
Specifications
Specifications
Polyester, Urethane Foam
PadMop™ Sponge Mop
Pharmaceutical production or aseptic processing areas, floors, walls, ceilings
18 in.
Documents