Micronova™ NovaHol™ Cleanroom Cleaner
This ready to use, fast drying, low-ionic detergent with added isopropyl alcohol is ideal for glass, acrylic and stainless steel cleanroom surfaces. Available in an easy to use, 1 quart spray bottle and a 1 gallon bottle.
Manufacturer:  Micronova™ NH1Q
 View more versions of this product
Catalog No. 19-130-0314
Description
Description
The Micronova NovaHol™ Cleanroom Cleaner is designed with a high evaporation rate, allowing direct application to a variety of surfaces including glass, acrylic, vinyl curtains, softwalls and stainless steel. The low concentration of Isopropyl Alcohol in the formula allows it to be used on vinyl and plastic surfaces.
  • Ready to use, cleanroom surfactant
  • Fast drying formula with added isopropyl alcohol
  • High evaporation rate
  • Ideal for conductive flooring
  • Works well with epoxy
  • Can be applied directly to a range of surfaces
  • Ideal for use on glass, acrylic and stainless surfaces in the cleanroom
  • Available in a 1 quart spray bottle and a 1 gallon refill bottle
  • Filtered to 0.1 microns
  • Low mobile ions
  • Low non-volatile residue
  • Biodegradable
  • Free-rinsing
Specifications
Specifications
Ideal for Glass, Acrylic, Vinyl curtains and Stainless Steel surfaces
1 qt.
0.1μm