Micronova™ M-Zone™ MittWipes™
Clean hard-to-reach areas easily and safely
$10.68 - $14.16
Specifications
For Use With (Application) Easily wipe around carts, shelving, tanks, equipment added protection for cage maintenance, cleaning, launderable and reusable
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Product Type Price Quantity  
Catalog Number Mfr. No. Includes Product Type Price Quantity  
19-142-609 Micronova™
MW201211
Foam interior Micronova Wipes Each for $14.16
Case of 24 Each for $300.24
19-142-608 Micronova™
MW2012
N/A Micronova Wipes Each for $10.68
Case of 24 Each for $226.56
Specifications
Description
  • Easily clean around carts, shelving, tanks, tubing and other hard-to-clean and hard-to-reach areas
  • Made with MegaTex fabric for heavily soiled or rough surfaces
  • Added protection in cleanroom maintenance, LAR applications
  • Autoclavable and launderable
  • Also available with foam interior for increased absorbency
MegaTex MittWipes with foam interior have increased absorbency.
Description & Specifications
Specifications
Easily wipe around carts, shelving, tanks, equipment added protection for cage maintenance, cleaning, launderable and reusable