Please login to your online account to display your discounted pricing

Micronova™ Labmarkers

With permanent, quick drying black ink that is resistant to IPA

$71.67 - $147.57

Specifications

Color Black
Type Labmarker
For Use With (Application) Marks on cold or wet surfaces, Plastic bags or disposable labware
Includes Fine tip
Quantity 10 Case
View More Specs
Products
Catalog Number Mfr. No. Format Price Quantity    

19-142-679

 
micronova™
PEN40
Standard Case of 10 Each for $71.67

19-142-678

 
micronova™
PEN40IR
Irradiated Case of 10 Each for $147.57
Description & Specifications

Specifications

Color Black
Type Labmarker
For Use With (Application) Marks on cold or wet surfaces, Plastic bags or disposable labware
Includes Fine tip
Quantity 10 Case

  • Fine tip
  • Use to mark on cold or wet surfaces, plastic bags or disposable labware
  • Will not smear on reusable glass, metal or porcelain labware but can be scrubbed off
  • Available in standard or gamma irradiated formats