Micronova™ Isolator Tool Adapters and Covers
Help to simplify cleaning in compliance with USP 797
$89.14 - $165.19
Specifications
For Use With (Equipment) Isolators, containment and bio hoods in cleanroom and buffer areas
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Product Type Price Quantity  
Catalog Number Mfr. No. Product Type Price Quantity  
19-435-082 Micronova™
QPIT09
Plastic isolator adapter tool Each for $89.14
19-160-0115 Micronova™
QDIT09
Mop Adapter Each for $165.19
Case of 6 Each for $904.98
Specifications
Description
  • Fit into small pass-thrus and docking devices yet allow extended reach
  • Two-ply polyester covers attach to isolator tool with adapter
Description & Specifications
Specifications
Isolators, containment and bio hoods in cleanroom and buffer areas