Please login to your online account to display your discounted pricing

Micronova™ CR100IR™ Irradiated Vinyl Cleanroom Tapes

Ultra-processed vinyl tape helps limit particulate and bioburden counts

$1,143.15 - $1,495.72

Specifications

Color White
Format Tape
Product Type Double bagged cleanroom Tape
For Use With (Application) Helps limit particulate and bio burden counts
Length (English) 36 yd.
View More Specs
Products
Catalog Number Mfr. No. Width (English) Width (Metric) Quantity Price Quantity    

19-306-980

 
micronova™
CR10034WHIR
0.75 in. 1.905cm 48 Case Case of 48 Each for $1,143.15

19-306-981

 
micronova™
CR10012WHIR
0.5 in. 1.27cm 72 Case Case of 72 Each for $1,495.72
Description & Specifications

Specifications

Color White
Format Tape
Product Type Double bagged cleanroom Tape
For Use With (Application) Helps limit particulate and bio burden counts
Length (English) 36 yd.
Length (Metric) 32.91m
Material Vinyl/Plastic

  • Wound on a plastic core and double bagged
  • Processed in a ISO 4 cleanroom environment
  • Useful for critical and aseptic processing applications