Please login to your online account to display your discounted pricing

Micronova™ Industrial-Grade Vinyl Cleanroom Tape

For light industrial use, clean construction and cleanroom support

$776.81 - $776.81

Specifications

Certifications/Compliance Complies with Federal Specifications PPP-T-66E and CID A-A 1689A
Format Tape
Sterile No
For Use With (Application) Aisle marking, labeling, batching, clean-build projects
Grade Industrial
View More Specs
Products
Catalog Number Mfr. No. Dimensions (L x W) Color Price Quantity    

19-149-323

 
micronova™
12WHEGVFP
0.5 in. x 36 yd. White Case of 72 Each for $776.81
Description & Specifications

Specifications

Certifications/Compliance Complies with Federal Specifications PPP-T-66E and CID A-A 1689A
Format Tape
Sterile No
For Use With (Application) Aisle marking, labeling, batching, clean-build projects
Grade Industrial
Material Vinyl

  • Designed for use in areas where ultraclean tape is not required
  • Forms a strong bond with plastics, polymers and stainless steel without leaving adhesive residue when removed
  • Can be written on with cleanroom pen
  • Processed in a controlled environment
  • Wound on a plastic core and sleeve-bagged
  • All rolls are 36 yd. long (33m)

Additional colors and widths available. Contact your Fisher Safety Customer Service Representative for ordering information.

Complies with Federal Specifications PPP-T-66E and CID A-A 1689A