Please login to your online account to display your discounted pricing

Micronova™ EGP™ Industrial-Grade Polyethylene Cleanroom Tape

Designed for use in cleanroom support areas, maintenance and clean build projects

$685.49 - $685.49

Specifications

Format Tape
Product Type Industrial grade cleanroom Tape
Quantity Each
Sterile No
For Use With (Application) Aisle marking, batching, color coding, clean-build projects, cleanroom support areas and food processing applications
View More Specs
Products
Catalog Number Mfr. No. Width (English) Color Price Quantity    

19-120-2532

 
micronova™
EGPFP1WH*
1 in. White Case of 36 Each for $685.49
Description & Specifications

Specifications

Format Tape
Product Type Industrial grade cleanroom Tape
Quantity Each
Sterile No
For Use With (Application) Aisle marking, batching, color coding, clean-build projects, cleanroom support areas and food processing applications
Grade Industrial
Length (Metric) 33m
Material Polyethylene

  • Used in areas where ultraclean tape is not required
  • Polyethylene backing with acrylic adhesive has low-outgassing and good clean-peel properties
  • Processed in controlled environment, wound on a plastic core, and sleeve bagged
  • Roll length: 33m (36 yd.)