Micronova™ Cleanroom Vinyl Tape
Processed for applications in and around cleanrooms
$18.72 - $52.56
Specifications
Certifications/Compliance FDA, CID
Format Tape
Product Type Cleanroom Tape
Quantity Each
Sterility Non-sterile
View More Specs
Products 12
Catalog Number Mfr. No. Color Width (English) Price Quantity  
Catalog Number Mfr. No. Color Width (English) Price Quantity  
19035333M View Documents Micronova™
12WHCR100PC
White 0.5 in. Each for $22.25
19035335M View Documents Micronova™
1WHCR100PC
White 1 in. Each for $26.74
Case of 36 Each for $857.16
19035337M View Documents Micronova™
2WHCR100PC
White 2 in. Each for $37.74
Case of 36 Each for $1,209.96
19035338M View Documents Micronova™
CR100PC3WH
White 3 in. Each for $52.56
Case of 36 Each for $1,684.80
191301254 View Documents Micronova™
15WHCR100PC
White 1.5 in. Each for $28.19
Case of 24 Each for $598.08
19-035-333F View Documents Micronova™
CR100PC12LG
Green 0.5 in. Each for $21.17
Case of 72 Each for $1,347.84
19-035-333G View Documents Micronova™
CR100PC12OR
Orange 0.5 in. Each for $21.64
Case of 72 Each for $1,387.44
19-035-333J View Documents Micronova™
CR100PC12RE
Red 0.5 in. Each for $21.17
Case of 72 Each for $1,347.84
19-035-333N View Documents Micronova™
CR100PC12YE
Yellow 0.5 in. Each for $18.72
Case of 72 Each for $1,273.68
19-035-333P View Documents Micronova™
CR100PC2LB
Light Blue 0.5 in. Each for $34.91
Case of 24 Each for $740.64
19-035-333R View Documents Micronova™
CR100PC2LG
Light Green 0.5 in. Each for $34.91
Case of 24 Each for $740.64
19-035-338 View Documents Micronova™
CR100PC34WH
White 0.75 in. Each for $24.56
Case of 48 Each for $1,049.76
Description
Description
Processed for applications in and around cleanrooms
  • Made of vinyl with synthetic rubber adhesive that ensures clean peel from wafer cartridges and most cleanroom work surfaces
  • Tapes and seals curved and irregular surfaces
  • Matte finish can be written on with cleanroom pen
  • Individually packaged in polyethylene bag
Order Info
Available in red, yellow, white, black, brown, dark blue, dark green, light blue, light green, orange, purple, and transparent in 36 yd. long (33m) rolls. Contact your Fisher Safety Customer Service Representative at 1-800-772-6733 for ordering information on colors not shown here.
Specifications
Specifications
FDA, CID
Cleanroom Tape
Non-sterile
Sealing, color coding, labeling, batching and food processing applications
33 m
Tape
Each
Matte
36 yd.
Synthetic Rubber,Vinyl