Please login to your online account to display your discounted pricing

Alfa Aesar™ Micronomicin sulfate, 590μg/mg

Chemical Identifiers

Chemical Name or Material Micronomicin sulfate
CAS 66803-19-8
Name Note 590μg/mg
Formula Weight 561.65
MDL Number MFCD00941451
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ6607209

  SDS
alfa aesar™
J6607209
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 561.65
Chemical Name or Material Micronomicin sulfate
CAS 66803-19-8
Physical Form Powder
MDL Number MFCD00941451
Color White to pale yellow
Storage Note 1 2° to 8°C
Quantity 10g
Name Note 590μg/mg
Solubility Information Freely soluble in water
Boiling Point 667°C
RTECS Number WK2196000

  • An aminoglycoside antibiotic commonly used against gram negative bacteria

Safety and Handling

2° to 8°C