Please login to your online account to display your discounted pricing
Fisher Scientific™ Micromaster™ Microscope Eyepiece Reticles
Adapt microscope eyepiece for counting, measuring or positioning
Products
Catalog Number Mfr. No. Price Quantity    
12-561-RG2
evos
MP-ERG-2
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications
  • Reticles mount inside 21mm I.D.
  • Micromaster I, II or III eyepieces, or 24mm Micromaster infinity eyepieces
  • Monocular Micromaster I, II or III models require focusable eyepiece 12-561-FEP