missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial, Cap and Septa Unassembled Component Kits with PTFE/Silicone/PTFE Septa DFS Item

Unassembled component kits offer the convenience of one product number for the vial, cap, and septa. Components are packaged to work with the unique MicroLiter Dispenser.

Manufacturer:  DWK Life Sciences 091040K

 View more versions of this product

Catalog No. 15-111-216



Description

Description

Available in multiple colors and septa formats and with or without writing patch.
Specifications

Specifications

Black
9mm
Limited Volume Vial Inserts - MicroLiter
PTFE/Silicone/PTFE
100
PTFE/Silicone/PTFE
12 mm
No
Polypropylene
Screw Thread
32mm
No
Vial Kit
Screw Thread
Borosilicate Glass (Type I)
2 mL
Documents
Promotions

Promotions