Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Microglobulin β -2 Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunofluorescence and flow cytometry experiments with the Thermo Scientific™ Microglobulin β-2 Ab-1, Mouse Monoclonal Antibody.

$240.59 - $847.63

Specifications

Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Murine
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS440P

 
thermo scientific™ lab vision™
MS440P
200μg/mL 1mL Each for $755.20

MS440P0

 
thermo scientific
MS440P0
200μg/mL 100μL Each for $240.59

MS440P1

 
thermo scientific™ lab vision™
MS440P1
200μg/mL 500μL Each for $602.55

MS440P1ABX

 
thermo scientific™ lab vision™
MS440P1ABX
1mg/mL 100μL Each for $680.04

MS440PABX

 
thermo scientific™ lab vision™
MS440PABX
1mg/mL 200μL Each for $847.63
Description & Specifications

Specifications

Applications Immunofluorescence
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Host Species Murine
Regulatory Status RUO
Research Discipline Hematopoietic
Type Primary
Immunogen Human PBL (T-ALL patient)
Isotype IgG2a

Ensure accurate, reproducible results in immunofluorescence and flow cytometry experiments with the Thermo Scientific™ Microglobulin β-2 Ab-1, Mouse Monoclonal Antibody.

Β2-microglobulin non-covalently associates with the 44 kDa α chain to form the HLA Class I antigen complex. Human Β2-microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. Clone HLA-ABC-m2 (also known as 246-E8.E7) is specific to human Β-microglobulin and does not react with primate cells.

Host Species: Mouse

Clone: 246-E8.E7; same as HLA-ABC-m2

Isotype: IgG2a

Species Reactivity: Human. Does not react with primate cells. Others not tested.

Epitope: Not determined

Immunogen: Human PBL (T-ALL patient)

Molecular Weight: 12kDa

Positive Control: Jurkat cells

Cellular Localization: Membrane

Recommended for:

  • Immunofluorescence
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO