Please login to your online account to display your discounted pricing
Microflex™ Tech One™ Latex Exam Gloves
$163.49 - $163.49
Products
Catalog Number Mfr. No. Price Quantity    
11-462-66B
microflex corporation
MF101S
Case of 1000 Each for $163.49
Description & Specifications
Lightly powdered with absorbable USP cornstarch for easier donning. Smooth glove surface provides a more natural feel. Made of high-quality, low-modulus latex for greater comfort. Beaded cuffs. Nonsterile. Ambidextrous.
Pack of 100 gloves. Length, 9 3/4 in. (24.8cm). Thickness, 7.5mil at fingertip.