Please login to your online account to display your discounted pricing
Microflex™ Nitron One™ Nitrile Exam Gloves
Lightly powdered with absorbable USP-grade cornstarch for easier donning
$16.89 - $174.42
Specifications
Coating Material Textured
Color Blue
Length (English) 9.6 in.
Length (Metric) 24.38cm
Material Nitrile
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    
19-048-722C
microflex corporation
NO-123-L
Large Pack of 100 Pair for $16.89
Case of 10 Pack for $161.50
19-048-722D
microflex corporation
NO-123-XL
X-Large Case of 1000 Pair for $174.42
19-048-722A
microflex corporation
NO-123-S
Small Pack of 100 Pair for $16.89
Case of 10 Pack for $161.50
19-048-722B
microflex corporation
NO-123-M
Medium Pack of 100 Pair for $16.89
Case of 10 Pack for $161.50
Description & Specifications
Specifications
Coating Material Textured
Color Blue
Length (English) 9.6 in.
Length (Metric) 24.38cm
Material Nitrile
Sterile No
Disposable Yes
Glove Cuff Style Beaded
Hand Ambidextrous
Latex Free No
Powdered Yes
Tensile Strength 18
Thickness 5.5mil
  • Textured surface for secure grip
  • Not made from natural rubber latex
Overall length: 9.6 in. (24.5cm). Palm thickness, Palm: 5.5mil., Fingers: 7.1mil. Ambidextrous. Single-use. Beaded cuff. Sold 100 gloves per pack.
510K approved for medical use