Please login to your online account to display your discounted pricing

Microflex™ Micro One™ Latex Exam Gloves

Formulated for proper comfort, fit and feel

$12.58 - $127.78

Specifications

Certifications ASTM D 3578
Coating Material Smooth
Color White
Length (English) 9.5 in.
Length (Metric) 24.1cm
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

18-044471

 
microflex corporation
MO150L
Large Case of 1000 Each for $125.84

19-044468

 
microflex corporation
MO150XS*
X-Small Case of 1000 Each for $127.78

19-044469

 
microflex corporation
MO150S
Small Pack of 100 Each for $12.58
Case of 10 Pack for $117.00

19-044470

 
microflex medical corp
MO150M*
Medium Pack of 100 Each for $20.86
Case of 10 Pack for $193.90

19-044472

 
microflex corporation
MO150XL*
X-Large Case of 1000 Each for $122.48
Description & Specifications

Specifications

Certifications ASTM D 3578
Coating Material Smooth
Color White
Length (English) 9.5 in.
Length (Metric) 24.1cm
Material Latex
Sterile No
Disposable Yes
For Use With (Application) Exam
Glove Cuff Style Beaded
Hand Ambidextrous
Powdered Yes
Thickness 5.5mil

  • Lightly powdered examination gloves feature smooth, nontextured surface
  • Interior is coated with absorbable USP cornstarch
  • Test values measure below proposed Federal quality and performance guidelines for maximum residual powder and protein content
  • Thickness is 6.3mil. minimum at fingertip
  • Beaded cuffs
  • Nonsterile
  • Ambidextrous
  • Single-use

100 gloves per pack. Length, 9.6 in. (24.5cm).

510K approved for medical use

Warning: These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.