Ansell MICROFLEX™ Integra™ EC N87 Nitrile Gloves
Nitrile gloves with extended cuff for added wrist and forearm protection are thick and durable to protect within a variety of healthcare, laboratory and industrial applications.
$3.31 - $17.54
Specifications
Certifications/Compliance AQL 1.5, FDA, ASTM F 1671, ISO9001
Color Blue
Disposable Single-use
Latex Free Yes
Material Nitrile
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-167-042
View Documents Promotion Details
Ansell
N871
Small
Pack of 50 Each for $17.54
Case of 10 Pack for $175.40
19-167-044
View Documents Promotion Details
Ansell
N873
Large
Pack of 50 Each for $17.54
Case of 10 Pack for $175.40
19-167-045
View Documents Promotion Details
Ansell
N874
X-Large
Pack of 50 Each for $4.97
Case of 10 Pack for $49.70
19-167-046
View Documents Promotion Details
Ansell
N875
2X-Large
Pack of 50 Each for $3.31
Case of 10 Pack for $33.10
Description
Description
Nitrile gloves with extended cuff for added wrist and forearm protection are thick and durable to protect within a variety of healthcare, laboratory and industrial applications.
Extra thickness offers great protection against bloodborne pathogens.
  • Powder free
  • Length: 12 in.
  • Palm: 8.0 mil
  • Medical/exam grade
  • Light blue
Specifications
Specifications
AQL 1.5, FDA, ASTM F 1671, ISO9001
Single-use
Nitrile
8 mil
21 MPa
30.5 cm
Blue
Yes
Powder Free
0.20 mm
12 in.
SDS
missing translation for 'documents'
missing translation for 'documents'