missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Microcosmic Salt Solution, LabChem™

$92.57 - $92.57

Chemical Identifiers

CAS 7732-18-5,7783-13-3
CAS Max % 95,5
Linear Formula H13NNaO8P
Formula Weight 209.07
Chemical Name or Material Microcosmic Salt Solution
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC169811 Lab Chem
LC169811
500mL Each for $92.57
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7783-13-3
H13NNaO8P
Microcosmic Salt Solution
95,5
209.07

Specifications

7732-18-5,7783-13-3
Liquid
209.07
Passes Test
Soluble in water
Room Temperature
Clear, colorless
Microcosmic Salt Solution
95,5
H13NNaO8P
1g/mL
Poly Bottle
1g/mL
Gaseous Ammonia; Phosphorus oxides
500mL

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.