Please login to your online account to display your discounted pricing
Thermo Scientific MICROCEN CL17 W/ RTR 120V
Quickly perform complex or simple protocols. Productivity, versatility, safety and convenience in a compact, easy to use lab instrument.
$3,697.13 - $3,697.13
Certifications/Compliance IEC 61010-1, IEC 61010-2, IEC 61010-020. CSA certified, CE marked, IVD compliant, Certified Biosafety
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Products
Catalog Number Mfr. No. Description Includes Capacity Voltage Price Quantity    
05-375-100
thermo sci centrifu bench
75002451
MicroCL 17 Centrifuge, Ventilated 24 x 1.5/2.0mL rotor with ClickSeal Biocontainment Lid 24 x 1.5/2mL 120V Each for $3,697.13
Description & Specifications
Specifications
Certifications/Compliance IEC 61010-1, IEC 61010-2, IEC 61010-020. CSA certified, CE marked, IVD compliant, Certified Biosafety
Capacity 24 x 1.5/2mL
Noise Level <56dbA
Timer 1 to 99 min.
Quickly perform complex or simple protocols with Thermo Scientific™ MicroCL 17 and 21 Microcentrifuges. When coupled with our extensive rotor selection, they offer productivity, versatility, safety and convenience in a compact, easy to use lab instrument.
Features:
  • Exceptional acceleration up to 21,000 × g in 12 seconds
  • Refrigerated versions offer cooling in just 9 minutes
  • Thermo Scientific™ ClickSeal™ biocontainment rotor lid “clicks” in place, ensuring securely contained samples
  • New PPSU lids have improved strength and chemical resistance
  • Intuitive controls and easy to read display simplifies operation
  • Rugged design for years of dependable service
  • Easy to clean and maintain
Rotor Portfolio:
  • 24 × 1.5/2.0mL Rotor with ClickSeal Biocontainment Lid certified by CAMR
  • 36 × 0.5mL rotor with screw-on lid
  • Dual Row rotor 18 × 2.0 /0.5mL with screw-on lid
  • PCR 8 × 8 rotor with screw-on lid
  • PCR 4 × 8 rotor with ClickSeal biocontainment lid
  • Hematocrit Rotor
  • Choice of two RCF options (17,000 × g/21,100 × g)
  • Refrigerated models (17R, 21R) offer temperature control from -9° to +40°C
Ordering Information:
Biocontainment, hematocrit, PCR and dual-row rotors, lids and tube adapters are available separately.