Please login to your online account to display your discounted pricing

Microbiology Wall Charts


The microscopic world of pathogens.

Manufacturer: fisher science education  WC54

 View more versions of this product

Catalog No. S67841

  • / Each
 

Description & Specifications

Specifications

Product Type Microbial Food Poisoning
Classification Science
For Use With (Application) Introduce students to the microscopic world of pathogens that cause microbial food poisoning