Microbe Scavenger Hunt
Explore the microworld in your environment.
Manufacturer:  Fisher Scientific AZ201075
 View more versions of this product
Catalog No. S32007
Includes:
  • Petri dishes with “ready-to-pour” media
  • “Microlife traps” for soil and water
  • Inoculating loops
  • Microscope slide and cover slips
  • Stain set
  • Pipets
  • Dichotomous key to molds, bacteria and protists
  • Guide to appropriate viewing techniques
  • Comprehensive teacher and student guides
Requires : Compound microscope
Description
Description
Hunt for various kinds of microorganisms — including bacteria, fungi and protists — in water, air, soil and even food, employing a ready-to-use growth medium.
  • Utilizes a specially formulated gel, which requires no autoclaving or special equipment
  • Makes it easy for students to design experiments that explore the subvisible world around them by collecting and identifying a wide variety of microbes
Order Info
Class size: 40 students.
Specifications
Specifications
Hunt for various kinds of microorganisms
Refill
Micro Scavenger Hunt Refill Kit
Education
Documents