Please login to your online account to display your discounted pricing

Microbe Scavenger Hunt


Explore the microworld in your environment.

Manufacturer: fisher science education  AZ-20-1073

 View more versions of this product

Compound microscope

Catalog No. S32006

  • / Each
 

Description & Specifications

Specifications

Class Size 40 students
Includes Petri dishes with ready to pour media, microlife traps for soil and water, inoculating loops, microscope slide and cover slips, stain set, pipets, Dichotomous key to molds, bacteria and protists, guide to appropriate viewing techniques, comprehensive teacher and student guides
For Use With (Application) Hunt for various kinds of microorganisms
Product Type Lab Investigation Kit