Micro Burners
Designed for micro-methods of analysis.
Manufacturer:  Fisher Science Education HUMH5815E
 View more versions of this product
Catalog No. S49118
Specifications Description & Specifications
Specifications
0.44 in.
1.1cm
Designed for micro-methods of analysis
3.5 in.
Nickel-plated, diecast zinc base, aluminum mixing tube and serrated gas inlet connection, adjustable gas valve
Micro Burner
0.31 in.
0.79cm
2000 to 3200 cu. ft.
8.7cm
LPG