Please login to your online account to display your discounted pricing

Miconazole (Free Base), Fisher BioReagents


Quantity 5g
Packaging Amber Glass

Manufacturer: fisher scientific company  BP26685

 View more versions of this product

Catalog No. BP26685

  • / Each
  • / Case of 27 Each
 

Description & Specifications

Chemical Identifiers

Melting Point 135°C
Formula Weight 416.13
Quantity 5g
Merck Index 15, 6257
Molecular Formula C18H14Cl4N2O
Physical Form Solid
Color White
Identification Pass Test
Purity Inclusive between 98.0%
Residual Solvents Pass Test
Health Hazard 2 WARNING!
Chemical Name or Material Miconazole
ChemAlert Storage Symbol Gray
CAS 22916-47-8
Recommended Storage RT
Packaging Amber Glass
Ignition Residue < 0.2%
Loss on Drying < 0.5%
Assay Percent Range 98%
Synonym 1-[2, 4-Dichloro-β-([2, 4-dichloro-benzyl]oxy)phenethyl]imidazole
Health Hazard 3 Emergency Overview
Harmful if swallowed. May cause allergic respiratory reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0
Name Note Free Base

Specifications

Chemical Name or Material Miconazole
Name Note Free Base
CAS 22916-47-8
Assay Percent Range 98%
Molecular Formula C18H14Cl4N2O
Formula Weight 416.13
Synonym 1-[2, 4-Dichloro-β-([2, 4-dichloro-benzyl]oxy)phenethyl]imidazole
Accessories
 

Emergency Overview
Harmful if swallowed. May cause allergic respiratory reaction. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. Get medical attention immediately if symptoms occur. Do not induce vomiting.

NFPA

Health:2
Flammability:0
Instability:0

WARNING!

RT

SDS & Safety Documents