missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Michler's Ketone, 98%

Manufacturer:  Alfa Aesar™ A1188922

 View more versions of this product

Catalog No. AAA1188922


 

Specifications

Specifications

Michler's Ketone
98%
MFCD00008312
14,6177
VVBLNCFGVYUYGU-UHFFFAOYSA-N
CN(C)C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)N(C)C
>360°C
7031
CHEBI:82347
268.36
90-94-8
C17H20N2O
790733
4,4'-Bis(dimethylamino)benzophenone
174°C to 176°C
bis[4-(dimethylamino)phenyl]methanone
268.36
100g
220°C (428°F)

Chemical Identifiers

90-94-8
268.36
VVBLNCFGVYUYGU-UHFFFAOYSA-N
7031
C17H20N2O
MFCD00008312
4,4'-Bis(dimethylamino)benzophenone
CHEBI:82347
Safety and Handling

Safety and Handling

GHS H Statement
H350-H341-H318
May cause cancer.
Suspected of causing genetic defects.
Causes serious eye damage.

GHS P Statement
P280-P281-P305+P351+P338-P310-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : DJ0250000

TSCA : TSCA

SDS Documents