Mica, Water Ground, 325 Mesh, Spectrum™ Chemical
M1289, 12001-26-2
$246.91 - $3815.18
Chemical Identifiers
CAS 12001-26-2
View More Specs
Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18604574
View Documents Promotion Details
Spectrum Chemical Mfg Cor
M1289500GM
500 g Poly Pail
Each for $246.91
18604575
View Documents Promotion Details
Spectrum Chemical Mfg Cor
M1289500GMCS6
6 x 500 g Poly Pail
Case of 6 Each for $905.74
18604577
View Documents Promotion Details
Spectrum Chemical Mfg Cor
M128925KGCS4
4 x 2.5 kg Poly Pail
Case of 4 Each for $1,979.38
18604578
View Documents Promotion Details
Spectrum Chemical Mfg Cor
M128945KGBL
45 kg Fiber Drum
Each for $3,815.18
Description
Description
Spectrum™ Chemical Mica, Water Ground, 325 Mesh is a wet-ground mica, a type of highly brittle silicate minerals with diverse chemical composition, that retains the brilliancy of its cleavage faces. It is used in cosmetic and personal care products as a colorant with opacifying properties. Its reflective and refractive properties make mica an important ingredient in makeup, moisturizing lotions, toothpastes, and nail polish. It appears as a clear, lustrous, crystalline solid plates and is insoluble in cold water.
Specifications
Chemical Identifiers
12001-26-2
Specifications
Poly Pail
500 g
Reagent
12001-26-2
100%
SDS
missing translation for 'documents'
missing translation for 'documents'