missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Invitrogen™ Human MICA Synthetic Peptide
Synthetic Peptide
Supplier: Invitrogen™ PEP0510
Description
MICA Synthetic Peptide, PEP-0510, from Invitrogen.
This peptide corresponds to 16 amino acids near the carboxy terminus of human MICA.PEP-0510 can be used as a blocking peptide with polyclonal antibody PA5-20393.Specifications
100507436 | |
purified | |
Human MICA | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
Q29983 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
MICA | |
0.02% sodium azide | |
50 μg | |
PBS |