missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MHC II (HLA-DR) Ab-1, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry experiments with the Thermo Scientific™ MHC II (HLA-DR) Ab-1, Mouse Monoclonal Antibody.

$245.35 - $846.65

Specifications

Antigen MHC II (HLA-DR) Ab-1
Clone LN3
Applications Flow Cytometry,Immunohistochemistry (Paraffin),Flow Cytometry,Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unlabeled
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
Catalog Number Mfr. No. Concentration Quantity Price Quantity  
MS133P Thermo Scientific™ Lab Vision™
MS133P
200μg/mL 1mL Each for $753.02
MS133P0 Thermo Scientific™ Lab Vision™
MS133P0
200μg/mL 100μL Each for $245.35
MS133P1 Thermo Scientific™ Lab Vision™
MS133P1
200μg/mL 500μL Each for $586.06
MS133P1ABX Thermo Scientific™ Lab Vision™
MS133P1ABX
1mg/mL 100μL Each for $674.29
MS133PABX Thermo Scientific™ Lab Vision™
MS133PABX
1mg/mL 200μL Each for $846.65
Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry and flow cytometry experiments with the Thermo Scientific™ MHC II (HLA-DR) Ab-1, Mouse Monoclonal Antibody.

HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of an α chain (36 kDa) and a β chain (27 kDa). It is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides and present them to T-helper cells. They therefore have a critical role in the initiation of the immune response.

Host Species: Mouse

Clone: LN3

Isotype: IgG2b

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Activated human peripheral blood mononuclear cells

Molecular Weight: 36kDa (α-chain) and 27kDa (β-chain)

Positive Control: Tonsil/lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

MHC II (HLA-DR) Ab-1
Flow Cytometry,Immunohistochemistry (Paraffin),Flow Cytometry,Immunohistochemistry (Paraffin)
Unlabeled
Concentrated
RUO
Primary
IgG2b
LN3
Monoclonal
Human
Murine
Hematopoietic
Activated human peripheral blood mononuclear cells