Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MHC I (HLA-A, B, C) Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MHC I (HLA-A, B, C) Ab-3, Mouse Monoclonal Antibody.

$251.73 - $860.33

Specifications

Antigen MHC I (HLA-A, B, C) Ab-3
Clone W6/32
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

MS1218P

 
thermo scientific™ lab vision™
MS1218P
200μg/mL 1mL Each for $772.53

MS1218P0

 
thermo scientific™ lab vision™
MS1218P0
200μg/mL 100µL Each for $251.73

MS1218P1

 
thermo scientific
MS1218P1
200μg/mL 500µL Each for $604.08

MS1218P1ABX

 
thermo scientific™ lab vision™
MS1218P1ABX
1mg/mL 100µL Each for $697.98

MS1218PABX

 
thermo scientific™ lab vision™
MS1218PABX
1mg/mL 200µL Each for $860.33
Description & Specifications

Specifications

Antigen MHC I (HLA-A, B, C) Ab-3
Clone W6/32
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Mouse
Regulatory Status RUO
Research Discipline Hematopoietic
Type Primary
Immunogen Cell membranes of human tonsil lymphocytes
Isotype IgG2a

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MHC I (HLA-A, B, C) Ab-3, Mouse Monoclonal Antibody.

Human MHC class I antigens (HLA-A, B and C) are expressed constitutively on all nucleated cells lymphocytes such as lymphocytes, thymocytes, granulocytes, and bone marrow cells and are absent on erythrocytes. MHC class I antigens play a role in class I MHC- associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation.

Host Species: Mouse

Clone: W6/32

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Cell membranes of human tonsil lymphocytes

Positive Control: Human lymphocytes, Tonsil and lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only; acetone fixation recommended)

Note:

USA: RUO; Int'l: RUO