missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ MHC I (HLA-A, B, C) Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MHC I (HLA-A, B, C) Ab-3, Mouse Monoclonal Antibody.

Manufacturer:  Thermo Scientific™ Lab Vision™ MS1218PABX

 View more versions of this product

Catalog No. MS1218PABX


 

Specifications

Description

Ensure accurate, reproducible results in immunohistochemistry experiments with the Thermo Scientific™ MHC I (HLA-A, B, C) Ab-3, Mouse Monoclonal Antibody.

Human MHC class I antigens (HLA-A, B and C) are expressed constitutively on all nucleated cells lymphocytes such as lymphocytes, thymocytes, granulocytes, and bone marrow cells and are absent on erythrocytes. MHC class I antigens play a role in class I MHC- associated antigen presentation, inhibition of NK cell cytotoxicity, tumor surveillance, and tissue allotransplantation.

Host Species: Mouse

Clone: W6/32

Isotype: IgG2a

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Cell membranes of human tonsil lymphocytes

Positive Control: Human lymphocytes, Tonsil and lymph node

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Frozen only; acetone fixation recommended)

Note:

USA: RUO; Int'l: RUO

Description & Specifications

Specifications

MHC I (HLA-A, B, C) Ab-3
Monoclonal
1mg/mL
Concentrated
Cell membranes of human tonsil lymphocytes
200μL
Hematopoietic
Monoclonal
Immunohistochemistry (Frozen)
W6/32
Unlabeled
Murine
IgG2a
RUO
Primary
Human